Would you like to log in again?
Autoradiography Film
Supplier: Alkali Scientific
XR1810 XR1570
104028-040EA 126.14 USD InStock
104028-040 104027-696
Autoradiography Film
Autoradiography Films Films
Capture autoradiography images effectively with BioBlue-MR™ Autoradiography Film, providing high-quality results for research experiments.
  • Ideal for a wide range of electrophoretic and autoradiography film applications
  • Can be used with calcium tungstate or blue-emitting rare-earth screens
  • E×cellent sensitivity and contrast
  • Compatible with automatic processors and manual development methods
  • Especially useful with chemiluminescence, BioBlue-×R works well with 14C, 32C, 33P, 125I, and 35S
BioBlue-MR™ Autoradiography Film is a premium-quality, blue-sensitive film meticulously crafted to cater to a broad spectrum of electrophoretic and autoradiography applications. Engineered to deliver exceptional performance, this film is compatible with both calcium tungstate and blue-emitting rare-earth screens, ensuring versatility in usage. Boasting remarkable sensitivity and contrast characteristics, BioBlue-MR™ is coated on both sides for enhanced consistency and overall efficiency. Its cutting-edge design consistently yields sharp, luminous images that are easily discernible and meet the standards for publication. Particularly adept in chemiluminescence applications, this autoradiography film is adept at capturing the nuances of radioisotopes such as 14C, 32P, 33P, 125I, and 35S, making it an indispensable tool in research and laboratory settings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty