Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Malachite green oxalate BS

Supplier: Ambeed
Danger
Synonyms: Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalateN-[4-[[4-(Dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-methanaminium, ethanedioate, ethanedioate(2:2:1)Basic Green 4N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate

A1371080-25G
77566-728EA 15 USD InStock
77566-728
Malachite green oxalate BS
Malachite green oxalate
Caution: For research use only.


Formula: C₅₂H₅₄N₄O₁₂
MW: 927.02 g/mol
MDL Number: MFCD00011766
CAS Number: 2437-29-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty