Would you like to log in again?
Bovine Serum Albumin (BSA)
Supplier: MP Biomedicals
0216006980 0216006990 0216006991 0216006925 0216006910 0216006950
IC16006980EA 606.31 USD InStock
IC16006980 IC16006991 IC16006990 IC16006950 IC16006910 IC16006925
Bovine Serum Albumin (BSA)
Proteins and Peptides
Albumins are a group of acidic proteins which occur in the body fluids and tissues of mammals and in some plant seeds. Serum and plasma albumin is carbohydrate free and comprises 55-62% of the protein present. However, only about 40% of the total albumin in the body is in the circulating plasma at one time with the remainder being in extracellular spaces with which there is, in general, equilibration about every 24 hours.
  • Presentation: Cream Colored Powder
  • Isoelectric point (pl): pI in Water at 25°C: Endogenous material - 4.7, 4.9; Fatty acid depleted- 5.3
  • Albumins are readily soluble in water and can only be precipitated by high concentrations of neutral salts such as ammonium sulfate.
  • Moisture: ≤6%
MP offers this grade of albumin as a more highly purified product than our traditional True Cohn Fraction V Powder. Due to lower manufacturing costs, this grade is available at a substantial savings relative to our True Cohn Fraction V Powder.
Albumin is general blocking reagent in ELISA, as a nutrient in cell & microbial culture, it also stabilize some enzymes during digestion of DNA. It also used to determine the quantity of other proteins, by comparing an unknown quantity of protein to known amounts of BSA.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty