Would you like to log in again?
Conductivity Cells, YSI
Supplier: YSI
603253 603256 003252 603255 603254
66121-998EA 955.65 USD InStock
66121-998 66121-996 66121-966 66121-994 66121-992
Conductivity Cells, YSI
Electrodes Conductivity Sensors
For use with YSI 3200 series conductivity meters.
Conductivity cells have built-in thermistors for automatic temperature compensation. Configurations include dip, flow, and small volume cells. Minimum volume for fill cell (66121-996) is 5mL. Volume for flow cell (66121-998) is 30mL. Cells are calibrated according to IOML (International Organization of Legal Metrology) recommendations 56 and 68.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty