Would you like to log in again?
pHRESH Refillable Combination pH Electrodes with Flow-on-Demand, Beckman Coulter®
Supplier: Beckman Coulter
511110 511111 511110
BK511111EA 403.98 USD InStock
CABK511110 BK511111 BK511110
pHRESH Refillable Combination pH Electrodes with Flow-on-Demand, Beckman Coulter®
Electrodes pH Electrodes
These electrodes measure the pH level in highly viscous or dirty samples such as soil, sludges, or slurries.
  • Epoxy and Glass Available
  • Ideal for Use with TRIS Buffers
  • Diameter: 12 mm (glass and epoxy)
  • Length: 125 mm (glass), 130 mm (epoxy)
  • Clogging Prevention Function
To prevent clogging, users can renew the junction after each use by pressing the electrode cap, which flushes the junction with filling solution.
The electrodes are ideal with TRIS buffers. Epoxy electrodes include a protective epoxy body that extends over the glass measurement bulb. BNC connectors are included. See BK511469 for reference solution refill.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty