Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunclon™ Delta Culture Tubes, Sterile, Thermo Scientific
Supplier: Thermo Scientific
Nunclon™
146183 146183
25382-343CS 743.68 USD InStock
25382-343 CA25382-343
Nunclon™ Delta Culture Tubes, Sterile, Thermo Scientific
Tubes Culture Tubes
These polystyrene, round-bottom Nunclon™Δ treated culture tubes are for use with rotators, producing cell growth evenly along the full inner surface.
Tested and certified for reliable quality. Radiation sterilized.
Ordering information: Supplied with polyethylene screw caps. Packaged 100 per sleeve.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty