Would you like to log in again?
Orion™ ROSS™ Half-Cell Electrode, Thermo Scientific
Supplier: Thermo Scientific
Orion™
8101BNWP
10010-754EA 572.48 USD InStock
10010-754
Orion™ ROSS™ Half-Cell Electrode, Thermo Scientific
Electrodes pH Electrodes
Precise pH determinations for routine or research work.
  • Electrode provides stable readings to 0.01 pH in less than 30 s
  • pH range of 0 to 14
  • Works extreme case of samples varying from one another by 50 °C or more
  • Precise pH determinations for routine or research work
  • Drift is less than 0.002 pH per day; restandardization is minimized
Get results three to five times more precise than those obtained with conventional electrodes with the Orion™ 8101BNWP ROSS™ Half-Cell Electrode. Use with Orion™ ROSS™ reference electrode 800300 or 800500U.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty