Would you like to log in again?
Orion™ Combination pH Electrodes, Thermo Scientific
Supplier: Thermo Scientific
ORION®
9102BNWP 9156SC
34107-747EA 509.32 USD InStock
34107-747 CA10010-778
Orion™ Combination pH Electrodes, Thermo Scientific
Electrodes pH Electrodes
Full-range, general-purpose, refillable Ag/AgCl combination pH electrodes are for accurate QC and research work.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty