Would you like to log in again?
Orion™ Needle Tip Micro Combination pH Electrode, Thermo Scientific
Supplier: Thermo Scientific
Orion™
9863BN
23227-008EA 1700.17 USD InStock
23227-008
Orion™ Needle Tip Micro Combination pH Electrode, Thermo Scientific
Electrodes pH Electrodes
This bevel needle-tipped 16-gauge glass body combination electrode can be used to measure pH by piercing septa.
  • Made of very thin, delicate glass
  • Single junction Ag/AgCl
  • Refillable
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty