Would you like to log in again?
Orion™ Combination pH Electrodes, Hard-Nose Bulb, Thermo Scientific
Supplier: Thermo Scientific
ORION®
9104BNWP
10010-784EA 726.04 USD InStock
10010-784
Orion™ Combination pH Electrodes, Hard-Nose Bulb, Thermo Scientific
Electrodes pH Electrodes
This pH electrode is ideal for carrying out everyday lab work.
  • Includes silver/silver chloride references that are widely used because of their accuracy and broad temperature range
  • With waterproof connector
Delivery information: Supplied with 60 ml bottle of Orion electrode fill solution.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty