Would you like to log in again?
Biopur Safe-Lock Microcentrifuge Tubes, Eppendorf®
Supplier: Eppendorf
022600001 022600001 022600044 022600028 022600028 022600044
53511-998CS 93.37 USD InStock
CA53511-998 CA53511-996 53511-998 CA53511-997 53511-996 53511-997
Biopur Safe-Lock Microcentrifuge Tubes, Eppendorf®
Tubes Microcentrifuge Tubes
Ideal for high-speed centrifugation, high-temperature incubations, enzyme assays, cloning procedures.
To meet the highest demands of the clinical, pharmaceutical, cosmetic, food industries and others, the quality and purity grade 'Biopur' was defined. This purity is ensured by an elaborate, automated manufacturing process which safeguards the product against contamination with biological substances. Certified sterile and free of pyrogens, DNases, RNases, DNA (human and bacterial), ATP, and PCR inhibitors. Individually blistered or 'mini batches' in resealable boxes provide effective protection against soiling and contamination.
Constructed of the highest quality virgin polypropylene, Eppendorf® Safe-Lock Tubes protect samples with excellent centrifugation stability, perfect sealing properties, and prevention of sample evaporation. Biopur Safe-Lock Tubes withstand up to 22000×g.
Delivery information: Supplied with a Quality Assurance Card of testing parameters. Lot-specific certificates available upon request.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty