Would you like to log in again?
Calomel pHree Epoxy Combination pH Electrode, Gel-Filled, Beckman Coulter®
Supplier: Beckman Coulter
A51705 A51705
BKA51705EA 240 USD InStock
CABKA51705 BKA51705
Calomel pHree Epoxy Combination pH Electrode, Gel-Filled, Beckman Coulter®
Electrodes pH Electrodes
Calomel pHree electrode for biological applications is a mercury-free alternative to calomel electrodes.
  • Rugged Epoxy Body over Glass Bulb
  • Mercury-Free Alternative to Calomel Electrodes
  • Fewer Precautions than Calomel Electrodes
  • Meter Long & Integrated Cable
  • BNC Connector
A silver-ion trap acts as a third junction to prevent contamination of samples with reference solution.
Unique chemistry allows electrode to deliver comparable performance to calomel electrode with fewer precautions for handling and disposal. Rugged epoxy body extends over glass bulb for protection. It features a one-meter-long and integrated cable with a BNC connector.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty