Would you like to log in again?
Spear-Tipped Combination pH Electrode, Double Junction, Gel-Filled, Beckman Coulter®
Supplier: Beckman Coulter
A57185 A57185
BKA57185EA 327.47 USD InStock
BKA57185 CABKA57185
Spear-Tipped Combination pH Electrode, Double Junction, Gel-Filled, Beckman Coulter®
Electrodes pH Electrodes
This electrode is used to measure pH levels in applications where sample piercing is required.
  • Rugged Epoxy Body
  • Durable, Spear-Shaped Tip
  • Integrated Cable
  • BNC Connector
  • Double-Junction Design Prevents Contamination
It has a rugged epoxy body with a durable, glass-measuring, spear-shaped tip
The electrode features one-meter-long, integrated cable with a BNC connector.
A double-junction design provides protection from sample contamination.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty