Would you like to log in again?
Epoxy Combination pH Electrode, Refillable, Beckman Coulter®
Supplier: Beckman Coulter
A57186 A57186
BKA57186EA 274.26 USD InStock
BKA57186 CABKA57186
Epoxy Combination pH Electrode, Refillable, Beckman Coulter®
Electrodes pH Electrodes
Epoxy Combination pH Electrode is for general purpose pH measurements.
  • Application: General Purpose pH Measurement
  • One-Meter Cable with BNC Connector
  • Protective Epoxy Body
  • Refillable
The rugged epoxy body extends over glass bulb for protection.
The electrode features an integrated, one-meter cable with a BNC connector.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty