Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glass Half-Cell pH Electrode, Beckman Coulter®

Supplier: Beckman Coulter
A57189 A57189
BKA57189EA 218.33 USD InStock
BKA57189 CABKA57189
Glass Half-Cell pH Electrode, Beckman Coulter®
Electrodes pH Electrodes
When indicating elements must be physically separated from reference elements to reduce noise or when a different reference element is needed to precisely match the measured sample, these electrodes are perfect.

  • Integrated, One-Meter Cable Included
  • BNC Connector
  • Diameter: 12 mm
  • Length: 150
  • Protective Epoxy Body

They feature one-meter-long, integrated cable with a BNC connector.

The electrodes are 12 mm x 150 mm.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty