Would you like to log in again?
InLab® Power Combination pH Electrode, METTLER TOLEDO®
Supplier: Mettler-Toledo
InLab®
51343110 51343110
97001-620EA 810 USD InStock
97001-620 CA97001-620
InLab® Power Combination pH Electrode, METTLER TOLEDO®
Electrodes pH Electrodes
<bold>Electrode features integrated SteadyForce™ reference system with under-over pressure to guarantee electrode discharge and prevent contamination
  • Integrated SteadyForce™ reference system
  • Ceramic junction
  • Constant, controlled discharge
  • A41 body
  • Resistant to corrosives
Ceramic junction allows for constant and controlled discharge of electrolyte. Special A41 glass body is resistant to corrosive or organic samples and is ideal for high temperatures and biological media. S7 screw head connects with different cables, so electrode may be used with a wide range of meters.
</bold>
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty