Would you like to log in again?
Orion™ Ammonia Ion Selective Electrodes, Thermo Scientific
Supplier: Thermo Scientific
Orion™
9512BNWP
10010-804EA 1254.69 USD InStock
10010-804
Orion™ Ammonia Ion Selective Electrodes, Thermo Scientific
Electrodes ISE Electrodes
This standard gas-sensing ISE electrode is designed for quick, reliable and accurate measurements of dissolved ammonia in aqueous solutions.
  • Waterproof BNC connector
  • Can be used on any ISE or mV meter with a BNC connection
The ammonia combination electrode has the sensing and reference half-cells built into one electrode, which decreases the amount of required solutions and reduces waste.
Accessories information: Use Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes. Polishing strips help restore the performance of solid-state, half-cell, or combination ion selective electrodes. Electrode handles are used with plastic membrane combination or half-cell electrodes.
Ordering information: Electrode is supplied with 40 loose membranes, one reusable membrane cap, tweezers for handling the membranes and one bottle of electrode filling solution with dispensing cap.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty