Would you like to log in again?
Orion™ ATC Probes, Thermo Scientific
Supplier: Thermo Scientific
ORION®
917001
34104-233EA 1022.15 USD InStock
34104-233
Orion™ ATC Probes, Thermo Scientific
Electrodes pH Electrodes
ATC probe for general purpose applications
  • With dual Banana plug
  • Durable epoxy body
  • For use with older/legacy Orion™ meters
For aqueous samples, as well as intermittent use in methanol or ethanol.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty