Would you like to log in again?
Calpain-Glo Protease Assay, Promega
Supplier: Promega
Calpain-Glo®
G8502 G8501
PAG8501EA 491.16 USD InStock
PAG8501 PAG8502
Calpain-Glo Protease Assay, Promega
Assays Enzyme Assays
The Calpain-Glo Protease Assay is a homogeneous, luminescent assay that measures calpain 1 (µ) and 2 (m) activities
  • Homogeneous, Luminescent Assay that Measures Calpain 1 (micro) and 2 (m) Activities
  • Simple "add-mix-measure" protocol
  • Maximum sensitivity while enzyme is still active
  • 1,000x more sensitive than fluorometric assays
The Calpain-Glo Protease Assay is a homogeneous, luminescent assay that measures calpain 1 (u) and 2 (m) activities. Calpains are a family of calcium-activated cysteine proteases involved in cleaving a wide variety of proteins. Calpains modulate the biological activities of their substrates via limited proteolysis. The Calpain-Glo Protease Assay provides a succinyl, proluminescent calpain substrate, Suc-LLVY-aminoluciferin, in a buffer system optimized for calpain and luciferase activities. The addition of the calpain reagent in an add-mix-measure format results in calpain cleavage of the substrate and rapid development of a glow-type luminescent signal produced by the luciferase reaction. The signal is proportional to the amount of calpain activity present. The assay is designed for use with purified enzyme preparations.
It is up to 1000X more sensitive than traditional fluorometric assays because it avoids inherent fluorescent background signals. Maximum sensitivity can be reached in as little as 10 minutes, so the enzyme is still fully active. Results are linear over four logs of calpain concentration.
The assay provides a succinyl, proluminescent calpain substrate, Suc-LLVY-aminoluciferin, in a buffer system optimized for calpain and luciferin activities. The addition of the calpain reagent in an add-mix-measure format results in calpain cleavage of the substrate and rapid development of a luminescent signal produced by the luciferase reactions. The signal is proportional to the amount of calpain activity present. The assay is designed for use with purified enzyme preparations and can be used in a multiwell format.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty