Would you like to log in again?
Conductivity Cells, YSI 3400 Series
Supplier: YSI
603440 603402 099821 603403 099820 603401
23201-045EA 857.91 USD InStock
23201-045 23201-012 23201-078 23201-125 23201-103 23201-205
Conductivity Cells, YSI 3400 Series
Electrodes Conductivity Sensors
For use with YSI conductive instruments as well as with instruments of other manufacturers.
These precision cells contain platinized platinum-iridium electrodes. 23201-125 has nickel-rod electrodes. Conductivity cells require cell adapter 66121-968 (sold separately) when used with YSI Models 3100 and 3200 conductivity meters. Choose cells made from Pyrex® 7740 or ABS plastic.
Note: Cell constant values are calibrated within 1% of cell constant.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty