Would you like to log in again?
Infekta-Pak 2 Ambient Infectious Specimen Shipper, InfeKta
Supplier: InfeKta
Infekta-Pak 2 Infekta-Pak
INF-2
89030-508CS 291.89 USD InStock
89030-508
Infekta-Pak 2 Ambient Infectious Specimen Shipper, InfeKta
Mailers and Shippers
Combination package is ideal for shipping 50mL conical centrifuge tubes, and TB and other ambient infectious specimens.
Preprinted outer box helps reduce shipping/marking errors. Package components include: a 10.8Wx10.8Dx18.4Hcm (41/4x41/4x71/4") preprinted white outer shipping box; a 5.1dia.x12.7Lcm (2x5") white fiberboard mailing tube with a seamed metal bottom, crimped metal neck, and matching metal screw cap; and an inner 95kPa, 162mL (5.5oz.), 4.4dia.x12.1Lcm (13/4x43/4") HDPE plastic bottle with a lined plastic screw cap. The markings on the outer box include UN certification registration, Class 6.2 infectious substance label, "to" and "from" address, and direction arrows. The outer box design also features a finger-locking "B" flute insert to hold the mailing tube securely in place during transport. The inner plastic bottle can accommodate 50mL conical centrifuge tubes.
Ordering information: A shipper's declaration form (not included) is required and must accompany the shipment of these packages.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty