Would you like to log in again?
Infekta-Pak 3 Ambient Infectious Specimen Shipper, InfeKta
Supplier: InfeKta
Infekta-Pak Infekta-Pak 3
INF-3
89030-510CS 269.13 USD InStock
89030-510
Infekta-Pak 3 Ambient Infectious Specimen Shipper, InfeKta
Mailers and Shippers
Combination package for the shipment of ambient infectious specimens of all risk groups.
Shipper components include a 10.8Wx10.8Dx28.6Hcm (41/4x41/4x111/4") white preprinted outer shipping box, a 9.5Wx9.5Dx16.5Hcm (33/4x33/4x61/2") "B" flute interlocking corrugated insert, a 7.9I.D.x28.3Hcm (31/8x111/8") kraft brown spiral-wound fiberboard mailing tube, and a 19.1Wx27.9Lcm (71/2x11") 7mil printed specimen bag with absorbent inner lining. Each outer shipping box is printed with UN certification registration for Class 6.2 infectious substance, "to" and "from" address, and directional arrows on two opposite box sides. Each mailing tube features a seamed metal bottom and crimped metal threaded neck (alpha thread only) with matching threaded cap. The inner 95kPa bag is printed with a black and orange biohazard symbol, instructions for use, and absorbent material laminated to the inside walls.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty