Would you like to log in again?
Orion™ Double Junction Gel-Filled pH Electrode, Epoxy Body, Thermo Scientific
Supplier: Thermo Scientific
ORION®
9156DJWP
14011-004EA 389.74 USD InStock
14011-004
Orion™ Double Junction Gel-Filled pH Electrode, Epoxy Body, Thermo Scientific
Electrodes pH Electrodes
This double junction pH electrode features a durable epoxy body.
  • With waterproof BNC connector
  • Low-maintenance gel filling
Low maintenance, non-refillable, epoxy body pH electrode with waterproof BNC connector. For use in TRIS buffers and with proteins. Works with any pH meter with a BNC connector. Durable. Mercury-free and environmentally safe.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty