Would you like to log in again?
Axygen® Hydrophobic Filter Barrier Pipette Tips, Corning
Supplier: Corning
Axygen®
TF-20-R-S TXLF-10 TF-1005-WB-R-S TXLF-10-R-S TF-50-R-S TF-205-WB-R-S TF-420-R-S TF-100-R-S TF-1000-R-S TF-400-R-S TF-150-R-S MTXF-1250-C-R-S TF-300-R-S TF-200-R-S
47745-490CS 1385.06 USD InStock
47745-490 47749-872 47745-492 10011-360 10011-370 10011-350 10011-364 10011-352 10011-354 10011-356 10011-358 12000-748 80082-898 80082-900
Axygen® Hydrophobic Filter Barrier Pipette Tips, Corning
Pipette Tips
Filtered tips are ideal for molecular and microbiology applications in which carryover contamination can invalidate results.
  • Tips are metal-free
  • Filters are made from polyethylene and contain no PCR inhibitors
Tips are designed to fit a wide variety of single-channel and multichannel pipettors. Flexible walls and a series of internal sealing rings ensure a secure fit and require less force to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.
All tips, except Extended Length MicroVolume Tips For Use With Most Research-Grade Pipettors bulk pack, are presterilized by e-beam radiation for protection from contamination.
The filter position allows for sufficient space between the sample and filter at the maximum rated volume and in case of overpipetting, the sample can be easily recovered by dispensing.
Certifications: Certified RNase-, DNase-, and endotoxin-free.
Ordering information: To order non sterile tips, contact your sales representative.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty