Would you like to log in again?
Axygen® 384-Well Rigi-Plate PCR Microplates, Corning
Supplier: Corning
Axygen®
PCR-384-RGD-C PCR-384-RGD-BK
89005-556CS 1140.06 USD InStock
89005-556 89040-032
Axygen® 384-Well Rigi-Plate PCR Microplates, Corning
Microplates PCR Plates
Flat bottom, deep well microplates are designed to provide extra rigidity in thermal cycling applications
Constructed of sturdy and durable polypropylene, plates are available in three color styles for application specific procedures. Choice of clear, black, or white plates allow for colorimetric, fluorescent, or luminescent applications. Certified RNase-, DNase-, and pyrogen-free. Well volume: 40µL.
Ordering information: Clear plates are packaged 20 plates per pack, 5 packs per case. Black and white plates are packaged in sleeves of 20, with 5 sleeves per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty