Would you like to log in again?
Caspase Inhibitor Set I, Calbiochem®
Supplier: Calbiochem
235429-1SET
80051-248EA 710.91 USD InStock
80051-248 CA80051-248
Caspase Inhibitor Set I, Calbiochem®
Cell Culture Supplements and Additives
Set contains 1mg samples of three caspase inhibitors: Caspase-3 Inhibitor II, Z-DEVD-FMK (80051-378), Caspase-1 Inhibitor I, Ac-YVAD-CHO (80052-266), and Caspase-1 Inhibitor V, Z-VAD-FMK (80054-792)
Supplied with an informational insert.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty