Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Midostaurin ≥98% (by HPLC), Calbiochem®

Supplier: MilliporeSigma
Synonyms: PKC412

539648-1MG 539648-1MG
80017-392EA 285.71 USD InStock
80017-392 CA80017-392
Midostaurin ≥98% (by HPLC), Calbiochem®
Midostaurin
CGP 41 251, N-Benzoylstaurosporine

Tan solid. This cell-permeable staurosporine derivative displays antitumor properties, and acts as a broad-spectrum, reversible, and ATP-competitive inhibitor of PKC, PDGFR[beta], VEGFR2, Syk, PKC[eta], PKC[delta], Flk1, Flt3, Cdk1/B, PKA, c-Kit, c-Fgr, c-Src, VEGFR1, and EGFR. At higher concentrations, it also inhibits InsR and IGF1R. Shown to potentially inhibit the proliferation of Ba/F3 cells expressing constitutively active Flt3. Soluble in DMSO.

Packaged under inert gas. Protect from light.


Formula: C₃₅H₃₀N₄O₄
MW: 570.649 g/mol
MDL Number: MFCD12828879
CAS Number: 120685-11-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty