Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BAY 61-3606 hydrochloride hydrate ≥97% (by HPLC), Calbiochem®

Supplier: MilliporeSigma

Packaged under inert gas.

574714-2MG 574714-2MG
80017-408EA 227.34 USD InStock
80017-408 CA80017-408
BAY 61-3606 hydrochloride hydrate ≥97% (by HPLC), Calbiochem®
BAY 61-3606 hydrochloride hydrate
BAY 61-3606, 2-(7-(3,4-Dimethoxyphenyl)-imidazo[1,2-c]pyrimidin-5-ylamino)-nicotinamide, HCl

Yellow solid. A cell-permeable imidazopyrimidine compound that acts as a potent, ATP-competitive, reversible, and highly selective inhibitor of Syk tyrosine kinase activity with no inhibitory effect against Btk, Fyn, Itk, Lyn, and Src at concentrations as high as 4.7µM. Shown to inhibit Syk-mediated cellular functions in vitro and exhibit good oral bioavailability and in vivo effacacy in the treatment of various allergy and asthma conditions in rat models. Soluble in DMSO.

Packaged under inert gas. Protect from light.


Formula: C₂₀H₁₈N₆O₃·HCl·H₂O
MW: 462.9 g/mol
Storage Temperature: Refrigerator
CAS Number: 732938-37-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty