Would you like to log in again?
VWR® Macro Pipette Tips
Supplier: VWR
VWR®
1028-745-300 1028-860-300 1025-860-300 1029-860-300 1027-860-300 1025-740-300 1029-745-300 1029-740-300
89368-992CS 231.39 USD InStock
89368-992 82018-842 89087-530 82018-840 89368-994 89368-982 89368-996 89087-532
VWR® Macro Pipette Tips
Pipette Tips
These clear polypropylene pipette tips are free of RNase, DNase, and trace metals.
  • All tips are autoclavable
Tips are available for VWR, Biohit®, Eppendorf®, Gilson®, Nichiryo®, HTL, Labnet, and Pipetman® macro pipettors.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty