Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Sickle Cell Test Kits, ASI, Arlington Scientific

Supplier: Arlington Scientific
ASI™
200100 200025
80087-322EA 121.86 USD InStock
80087-322 80087-320
Sickle Cell Test Kits, ASI, Arlington Scientific
Clinical Diagnostic Supplies Clinical Reagents and Kits
The ASI Sickle Cell Test is a convenient and specific kit intended to be used as an aid in the qualitative detection of hemoglobin S (Hb-S) in anticoagulated whole blood. The test does not distinguish between sickle cell disease (HbS/S) and sickle cell trait (HbS/A). This test is not recommended for use on newborns under 3 months of age. Kit does not include controls.

  • Accurate, easy and economical
  • Detects both homozygous (S/S) and heterozygous (A/S) sickle cell
  • Contains urea reagent for test confirmation
  • Turbidity line test based upon modified Nalbandian procedure
  • Room temperature storage
  • Proudly made in USA
  • Longest shelf life - up to 18 Months
  • Sensitivity: 99%
  • Specificity: 99%

Certifications: CE Marked
CPT Code 85660
510(k)-K960947

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty