Would you like to log in again?
Accessories for PYREX® Continuous Liquid/Liquid Extractor, Heavier Than Water, One Piece, Corning
Supplier: Corning
PYREX®
3920-MBO
89091-652CS 556.49 USD InStock
89091-652
Accessories for PYREX® Continuous Liquid/Liquid Extractor, Heavier Than Water, One Piece, Corning
Extractors Liquid-Liquid Extractors
Made of PYREX® borosilicate glass. These continuous extractor systems are for use with solvents of heavier specific gravity than sample liquid being extracted.
  • Extraction can be performed outside the hood
  • Designed to meet EPA test method requirements outlined by SW-846, RCRA, and CLP
The sidearm is sealed to the extractor body just below the 45/50 [ST] outer joint. The sidearm is sealed higher on the body to reduce the chance of water spilling over while draining remaining solvent from the extractor body at the end of the extraction. Extraction can be performed outside the hood. However, as dictated by safe laboratory practices, it is recommended that some type of ventilation system be used above or near the units in case of a drop in condenser water temperature or loss of condenser cold water supply.
Delivery information: Supplied with Allihn condenser with 45/50 [ST] inner joint and 500m; PYREX® round bottom boiling flask with 24/40 [ST] outer joint.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty