Would you like to log in again?
P Series Plastic Tanks, Grant Instruments
Supplier: Grant
P18 P12
47727-838EA 586.8 USD InStock
47727-838 47727-836
P Series Plastic Tanks, Grant Instruments
Baths Water Baths, Unstirred
These economy tanks are constructed of rugged polyethylene plastic and double walled for rigidity
The curved plastic lids corresponding to the plastic tanks are required for operating a circulating water bath at elevated temperatures (60° to 99°C).
They provide excellent results for water usage up to 99°C with an operating range of 5° above ambient to 99°C. Tanks have a minimum liquid working depth of 8.3cm (31/4") and a maximum of 14cm (51/2").
Ordering information: All tanks are supplied with a bridge plate for mounting the circulator. Lids sold separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty