Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trichloroethylene 99.6% stabilized ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,1,2-TrichloroetheneTrichloroethene1,1,2-Trichloroethylene

421520025 421525000
TS421520025EA 247 USD InStock
TS421520025 TS421525000
Trichloroethylene 99.6% stabilized ACS
Trichloroethylene


Formula: ClCH=CCl₂
MW: 131.39 g/mol
Boiling Pt: 86.7 °C (1013 hPa)
Melting Pt: –85 °C
Density: 1.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000838
CAS Number: 79-01-6
UN: 1710
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.02 % (Coulometric)
GC ≥99.5 %
Color scale ≤10 APHA
Stabilizer ≤1 % (Type of stabilizer mentioned on the coa)
Residue after evaporation ≤0.001 %
Free halogens Passes test
Heavy metals (as Pb) ≤1 ppm
Titrable acid ≤0.0001 meq/g
Titrable base ≤0.0003 meq/g

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty