Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodic acid 99.6% ACS

Supplier: Thermo Scientific Chemicals
Danger

423811000 423815000
TS423811000EA 228 USD InStock
TS423811000 TS423815000
Iodic acid 99.6% ACS
Iodic acid


Formula: HIO₃
MW: 175.91 g/mol
Melting Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00011348
CAS Number: 7782-68-5
EINECS: 231-962-1
UN: 3085
ADR: 5.1,II
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder, crystals and chunks
Titration Iodometric ≥99.5 %
Residue after ignition ≤0.02 %
Chloride and Bromide ≤0.02 % (as Cl)
Iodide (I) passes test
Sulfate (SO4) ≤150 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %
Nitrogen compounds (as N) ≤0.1 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty