Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trichloroethylene 99+% stabilized, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,1,2-TrichloroetheneTrichloroethene1,1,2-Trichloroethylene

158310025 158310250
TS158310025EA 112 USD InStock
TS158310025 TS158310250
Trichloroethylene 99+% stabilized, Extra Pure
Trichloroethylene


Formula: ClCH=CCl₂
MW: 131.39 g/mol
Boiling Pt: 87 °C (760 mmHg)
Melting Pt: –85 °C
Density: 1.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000838
CAS Number: 79-01-6
EINECS: 201-167-4
UN: 1710
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4780 (20°C, 589 nm)
Water ≤0.015 % (Coulometric)
GC ≥99.0 %
Color scale ≤15 APHA
Stabilizer Stabilized
Residue after evaporation ≤0.002 %
Heavy metals (as Pb) 0.001 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty