Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitromethane-[D₃] 99.5 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Synonyms: Nitromethane-[²H₃]

464230050
TS464230050EA 95.1 USD InStock
TS464230050
Nitromethane-[D₃] 99.5 atom % D for NMR spectroscopy
Nitromethane-[D3]


Formula: D₃CNO₂
MW: 64.02 g/mol
MDL Number: MFCD00044214
CAS Number: 13031-32-8
UN: 1261
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty