Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
StatSpin® MP Multipurpose Centrifuge, HemoCue
Supplier: HemoCue
StatSpin®
SSMP SSMP
87002-248EA 4321.6 USD InStock
CA56430-006 87002-248
StatSpin® MP Multipurpose Centrifuge, HemoCue
Centrifuges Microcentrifuges
This small, high-speed centrifuge is designed to run a variety of applications quickly, efficiently, and quietly, making it an ideal general-purpose centrifuge for the stat lab, clinic, or office laboratory.
  • Low-Vibration Processing
  • Rotors Reach Top Speed in Seconds
  • Lightweight, Low-Mass Rotor
  • Easy Rotor Change
  • Ideal for Fingerstick Procedures
  • General-Purpose
The unique drive and suspension system results in nearly vibration-free operation
Small sample requirements make it ideal for fingerstick procedures or venous blood samples up to 3 mL.
Lightweight, low-mass rotors achieve both top speed and full breaking in a few seconds. Rotors can easily be changed with simple pull-off and push-on actions to transform the centrifuge from hematocrit to urine sediment to plasma applications.
Cycle time: 45 s urine, 120 s hematocrit, 30 s tube rotors, 30 s plasma rotor, 95 s lipoclear, 120 s coag plasma
Certifications: UL listed. CE marked.
Ordering information: Includes one-year manufacturer’s warranty, 1.5 mL tube rotor (87002-238), hematocrit rotor (87002-234) and circular tube reader, startup supply of PlasmaRotor® rotors, universal power supply, and 120V adapter and power cord. For lipemia clearing reagent and tubes, see 87002-124 and 87002-118 series. For microhematocrit tubes, see 87002-148 series.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty