Would you like to log in again?
KERLIX™ Super Sponges, Sterile, Covidien
Supplier: Covidien
KERLIX™
3085
82004-770CS 513.62 USD InStock
82004-770
KERLIX™ Super Sponges, Sterile, Covidien
First Aid Supplies Wound Dressings
Prewashed, fluff-dried woven gauze has diamond-fold construction and a crinkle-weave pattern to offer superior loft, bulk, and high absorbency.
This superior primary dressing is designed to transfer exudate away from the wound into the secondary dressing. Absorbency, strength, and low-linting characteristics are ideal for pre-operative prepping, cleansing, debriding, and packing. Sponges are made from high-grade 100% cotton.
Certifications: Exceed USP Type VII gauze requirements.
Ordering information: Supplied in soft pouches, five individually-wrapped sponges per pouch.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty