Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VISTEC® X-Ray Detectable Sponges, Covidien
Supplier: Covidien
VISTEC®
7148
82004-810CS 1068.66 USD InStock
82004-810
VISTEC® X-Ray Detectable Sponges, Covidien
First Aid Supplies Wound Dressings
Sponges have a unique dual-colored element that combines a blue radiopaque strand with a white strand, which is visible when the sponge is saturated with blood.
The element is latex-free and runs through both sides of the sponge to facilitate counting. Precise, double vacuum delinting process assures low lint.
Certifications: Meet USP Type VII gauze requirements.
Ordering information: All sponges are paper-banded in tens. Nonsterile sponges are packaged in bags of 100; sterile sponges are tray-packed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty