Would you like to log in again?
Yeast DNA Extraction Kit, Thermo Scientific
Supplier: Thermo Scientific
78870 78870
PI78870EA 253.66 USD InStock
PI78870 CAPI78870
Yeast DNA Extraction Kit, Thermo Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Extract and purify genomic and plasmid DNA from yeast in less than one hour using the materials provided in this kit.
  • Greater yields than traditional methods
  • Eliminates need for glass beads or harsh enzyme treatments
  • Yields DNA that is compatible with PCR amplification
  • Scalable to allow purification from single colonies up to 500 ml of cultured yeast
The kit uses Y-PER™ reagent to quickly and easily lyse yeast cells without requiring any enzymatic treatment of glass beads. The resulting DNA contains little or no RNA contamination and is ready to be used for PCR or transformation applications.
Delivery information: Kit includes Y-PER Yeast Protein Extraction Reagent (25 mL), DNA Releasing Agent A and B (20 mL of each), and Protein Removal Reagent (10 mL).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty