Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Penetration Needle, Humboldt
Supplier: Humboldt
H-1280
52934-120EA 689.26 USD InStock
52934-120
Penetration Needle, Humboldt
Penetrometer Needles
For penetration test of bituminous materials as specified in ASTM Method D5
Hardened stainless steel, with brass shank. Exposed needle length: 40–45mm (11/2–13/4").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty