Would you like to log in again?
SP Bel-Art Sterile Cloning Discs, Bel-Art Products, a part of SP
F37847-0002 F37847-0001 F37847-0003
13196-102PK 63.03 USD InStock
13196-102 13196-104 13196-106
SP Bel-Art Sterile Cloning Discs, Bel-Art Products, a part of SP
Nucleic Acid Storage Systems
Saves time and reduces effort when isolating a large number of clones.
  • Sterile
  • Simplify isolating a large number of clones
  • Quickly and easily transfer cells
The sterile paper discs transfer cells quickly and easily to a 24-well plate. Cloning discs have been sterilized by Gamma radiation. 2 vials of 50 discs per pack.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty