Would you like to log in again?
Ranger Surface-Mix Hand Torch, Wale Apparatus
Supplier: Wale Apparatus
90-9300
18363-005EA 802.55 USD InStock
18363-005
Ranger Surface-Mix Hand Torch, Wale Apparatus
Heaters Gas Burners
Provides a hot quiet flame without flashback
Lightweight stainless steel construction with wood handle and nickel-plated brass valve. Performs well with fuel pressures from 0.25 to 5 psi for natural gas and propane; from 0.25 to 15 psi for oxygen.
Ordering information: Order torch and twin-tubing connectors separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty