Would you like to log in again?
Comply™ SteriGage™ Sterilization Integrators, 3M™
Supplier: 3M
Comply™
MMM 1243A 7000002707
34010-019CS 273.82 USD InStock
34010-019 CA34010-019
Comply™ SteriGage™ Sterilization Integrators, 3M™
Sterilization Indicators
Provide a simple, accurate method of assuring that all conditions necessary for sterilization have been met during a sterilization cycle.
Steam sterilization integrator can be used in all steam sterilization processes, including vacuum, gravity, and flash sterilization. Integrates time, temperature, and steam just like live spore strips. Accurate to ±1 minute at 132 °C (270 °F). Works at all normal sterilization temperatures, eliminating multiple inventories.
Steam integrators are available with optional extender or recording card. Extender is ideal for verifying integrators without contaminating sterilized materials in large packs. Integrator plus extender is 23 cm (9³/₁₆") long. Recording card provides a permanent record of sterilizer use and work flow.
Integrators are held together with a temperature-resistant, pressure-sensitive adhesive. Information is displayed in a precise, easy-to-read manner to tell user whether to accept or reject the pack, or load immediately after the sterilization cycle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty