Would you like to log in again?
Standard Ball Valve Lockout, Brady®
Supplier: Brady
65666 65666
89011-764EA 65.25 USD InStock
CA89011-764 89011-764
Standard Ball Valve Lockout, Brady®
Security Locks
Ideal for securing valve handles in the on or off position
  • Easy to use, single-piece design
  • For use on ball valve sizes from 6-25mm (1/4-1")
  • Accepts 7mm (9/32") padlock shackles
Easy to use, single-piece design makes the standard ball valve lockouts the perfect tool for securing valve handles in the off position.
The hole diameter accepts 7.14mm (⁹⁄₃₂") padlock shackles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty