Would you like to log in again?
BCR® High Efficiency Mask, Berkshire
Supplier: Berkshire
BCR®
BM.1.12
21923-584CS 700.3 USD InStock
21923-584
BCR® High Efficiency Mask, Berkshire
Masks Cleanroom Masks
Three-layer mask for controlled environments.
  • Non-shedding
  • Polyurethane earloops reduce particulation
  • Headband option provides more secure facial seal
  • Ultrasonic bonding reduces contamination and outgassing
  • Cleanroom Class 1 - 100000
High efficiency (96% at 1 µm) polypropylene filter is held between 100% nylon outer layer and 100% rayon inner layer. Pressure differential of 1.98 for continuous comfort and breathability. Polyurethane ear loops can be worn around the ears or fastened with a polyethylene clasp. Ultrasonically bonded seams.
Certifications: ISO Class 3–8
Ordering information: Individually wrapped or bulk packaging options.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty