Would you like to log in again?
Thermocouple Vacuum Gauge, Welch®
Supplier: Gardner Denver
1515
55008-180EA 1488.24 USD InStock
55008-180
Thermocouple Vacuum Gauge, Welch®
Pumps Vacuum Gauges
This gauge is portable and lightweight (less than one kg)
Caution: Gauge is NOT to be used to measure the pressure of explosive gases or vapors.
It includes a 0 to 10V DC recorder output and has a range of 0 to 5000mtorr.
Ordering information: Supplied with one permanently attached probe cable, gauge tube, and carrying handle. Three-wire cord and plug included.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty