Would you like to log in again?
Weller® Controlled-Output Iron, Cooper Tools, Apex
Supplier: Apex
Weller®
WP35
37698-428EA 174.35 USD InStock
37698-428
Weller® Controlled-Output Iron, Cooper Tools, Apex
Soldering Supplies Soldering Irons
Versatile iron features three interchangeable heating elements (25, 30, and 35W) so that wattage can be changed for different applications
Additional tips are available in different styles and sizes, all designed to provide maximum heat transfer from heater to tip. Tips feature a copper core electroplated with iron to extend the lifetime of the tip. Nickel and chromium behind the working surface protect the shank from corrosion and solder creep. All tips are individually bagged.
Quick-change knurled collar makes changing elements simple. Heating elements are nichrome wound with fiberglass and ceramic insulation. Lightweight handle has cushion grip. Iron comes with screwdriver tip 37691-434, iron stand, and 1.8m (6') three-wire grounded power cord. Tip temperature: 454°C (850°F). Electrical: 120V, 35W.
Certifications: Iron is UL and C-UL listed.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty