Would you like to log in again?
Wall-Mounted Autopsy Stations, Mortech®
Supplier: Mortech
1036-7 1036-10 1036-8
29000-012EA 73403.88 USD InStock
29000-012 29000-010 29000-008
Wall-Mounted Autopsy Stations, Mortech®
Furniture Casework
All stainless steel stations feature heliarc-welded seams and joints that are ground polished to a smooth finish.
  • Plumbing and electrical lines are factory installed and require only minimal installation
  • All inside corners have large radii for easy cleaning and effective sanitation
  • The reverse flow feature instantly relieves suction clogging by providing a reverse pressure of water
  • Hot/cold water fixture features a gooseneck faucet and wrist-blade handles
Large, single compartment sinks feature a heavy-duty ¹/₂ hp commercial waste disposal. Sinks are 86.4 cm (34") wide and feature a deluxe aspirator and vacuum beaker with reverse flow for additional waste removal. A locking mechanism with a slide latch secures carts to the sink; mechanism can be accessed from either side of the carrier. Stations feature a 111.8 cm (44") wide dissecting area next to the sink and a 30.5 cm (12") deep specimen shelf on top. The dissecting area rinse assembly and the spray hose assembly feature cold water control valves. Spray hose assembly has a nozzle and 3 m (10') of flexible hose. Stations are equipped with two waterproof GFCI outlets and a waterproof switch for the waste disposal. A large instrument drawer is located below the dissecting area on the right and left sink models; center sink model features two dissecting areas and two instrument drawers.
Note: Specified dimensions reflect total height when unit is installed with countertop area 86.4 cm (34") above the floor.
Ordering information: Plumbing fixtures can be located in various positions; please specify requirements when ordering. Optional features such as eyewash assemblies, gas/air valves, and light fixtures are available. Contact your Avantor representative for more information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty