Would you like to log in again?
Mortuary Refrigerators, Mortech®
Supplier: Mortech
1036-R116 1036-R106 1036-R104
29000-044EA 59514.35 USD InStock
29000-044 29000-046 29000-042
Mortuary Refrigerators, Mortech®
Refrigerators
Refrigerators operate at 3°C (37°F) and are equipped with a condensing unit that is air-cooled with R22 refrigerant
The top, sides, and rear of each unit are constructed of 20-gauge stucco-embossed aluminum. The front is constructed of Type 304 stainless steel with a No. 4 finish. Doors close flush with the unit body and feature a magnetic gasket and a cardholder for identifying contents. The compartment interior features 20-gauge stucco-embossed aluminum for the walls and ceiling and 14-gauge smooth aluminum for the floor. Each compartment features a three-piece telescopic sliding carriage frame constructed of extruded aluminum. Carriage frames have nylon rollers for easy sliding. All hardware is chrome-plated. A lighted switch on the exterior controls an interior vapor-proof light and a dial thermometer on the front of the refrigerator monitors interior temperature. The electrical junction box is located on the top of the unit.
The two-body side acting refrigerator is ideal for areas with limited space in front of the unit. All units feature 10.2cm (4") foamed-in-place urethane insulation that is UL listed.
The two-body and three-body units feature 1/2hp condensing units; the four-body model features a 3/4hp condensing unit. Condensing units include a removable grill.
Ordering information: Refrigerators are supplied with one removable, 18-gauge Type 304 stainless steel tray for each compartment. For additional electrical configurations and refrigerants, contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty